missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Orotic Acid Monohydrate 98.0+%, TCI America™

$25.97 - $282.64

Chemical Identifiers

CAS 50887-69-9
Molecular Formula C5H6N2O5
Molecular Weight (g/mol) 174.112
MDL Number MFCD00006027
InChI Key YXUZGLGRBBHYFZ-UHFFFAOYSA-N
Synonym 2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
PubChem CID 16218504
IUPAC Name 2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
SMILES C1=C(NC(=O)NC1=O)C(=O)O.O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O006525G View Documents TCI America
O006525G
25g Each for $25.97
 
O0065100G View Documents TCI America
O0065100G
100g Each for $79.45
 
O0065500G View Documents TCI America
O0065500G
500g Each for $282.64
 
Specifications

Chemical Identifiers

50887-69-9
174.112
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
16218504
C1=C(NC(=O)NC1=O)C(=O)O.O
C5H6N2O5
MFCD00006027
2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate

Specifications

White
50887-69-9
MFCD00006027
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
16218504
Crystal-Powder at 20°C
Orotic Acid Monohydrate
25g
C5H6N2O5
2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid
C1=C(NC(=O)NC1=O)C(=O)O.O
174.112
156.10
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0278

RTECSNumber : RM3180000

TSCA : Yes