missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Orotic Acid Monohydrate 98.0+%, TCI America™

Manufacturer:  Tci America O006525G

 View more versions of this product

Catalog No. O006525G



Specifications

Specifications

OroticAcidMonohydrate
25g
C5H6N2O5
2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, orotc acd hydrate?, orotic acid hydrate, orotic acid monohydrate, oroticum acidum, uracil-4-carboxylic acid hydrate, uracil-6-carboxylic acid monohydrate
C1=C(NC(=O)NC1=O)C(=O)O.O
174.112
156.10
≥98.0% (HPLC,T)
White
50887-69-9
MFCD00006027
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
16218504
Crystal-Powder at 20°C

Chemical Identifiers

50887-69-9
174.112
YXUZGLGRBBHYFZ-UHFFFAOYSA-N
16218504
C1=C(NC(=O)NC1=O)C(=O)O.O
C5H6N2O5
MFCD00006027
2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid, 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate, lactinium, orotc acd hydrate?, orotic acid hydrate, orotic acid monohydrate, oroticum acidum, uracil-4-carboxylic acid hydrate, uracil-6-carboxylic acid monohydrate
2,4-dioxo-1H-pyrimidine-6-carboxylic acid;hydrate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0278

RTECSNumber : RM3180000

TSCA : Yes

Documents