Please login to your online account to display your discounted pricing

Orotic acid monohydrate, 98%, ACROS Organics™

$74.82 - $74.82

Chemical Identifiers

Chemical Name or Material Orotic acid monohydrate
CAS 50887-69-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4N2O4·H2O
Formula Weight 174.11
MDL Number MFCD00149398
Synonym 2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid monohydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC129571000

  SDS
fisher scientific acros
129571000
Plastic bottle 100g Each for $74.82

AC129570050

  SDS
thermo fisher scientific
129570050
Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC129575000

  SDS
thermo fisher scientific
AC129575000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 174.11
Quantity 100g
Color White to off-white
Beilstein 25, 253
Merck Index 15, 6972
Chemical Name or Material Orotic acid monohydrate
Assay Percent Range 98%
Molecular Formula C5H4N2O4·H2O
CAS 50887-69-9
Infrared Spectrum Authentic
Loss on Drying 12.0% max. (1 g, 130°C, 3 hrs)
MDL Number MFCD00149398
Packaging Plastic bottle
Synonym 2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid monohydrate
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.