Please login to your online account to display your discounted pricing

Orotic acid monohydrate, 98%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC129575000

 View more versions of this product

Catalog No. AC129575000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 174.11
Quantity 500g
Color White to off-white
Beilstein 25, 253
Merck Index 15, 6972
Loss on Drying 12.0% max. (1 g, 130°C, 3 hrs)
Synonym 2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid monohydrate
Molecular Formula C5H4N2O4·H2O
CAS 50887-69-9
Assay Percent Range 98%
Chemical Name or Material Orotic acid monohydrate
MDL Number MFCD00149398
Packaging Plastic bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Orotic acid monohydrate
CAS 50887-69-9
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H4N2O4·H2O
Formula Weight 174.11
MDL Number MFCD00149398
Synonym 2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid monohydrate
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents