Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody.

$280.10 - $1,246.84

Specifications

Antigen Ornithine Decarboxylase (ODC) Ab-1
Clone MP16-2
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS464P

 
thermo scientific™ lab vision™
MS464P
200μg/mL 1mL Each for $1,153.10

MS464P0

 
thermo scientific™ lab vision™
MS464P0
200μg/mL 100µL Each for $280.10

MS464P1

 
thermo scientific™ lab vision™
MS464P1
200μg/mL 500µL Each for $778.98

MS464P1ABX

 
thermo scientific™ lab vision™
MS464P1ABX
1mg/mL 100µL Each for $871.20

MS464PABX

 
thermo scientific™ lab vision™
MS464PABX
1mg/mL 200µL Each for $1,246.84
Description & Specifications

Specifications

Antigen Ornithine Decarboxylase (ODC) Ab-1
Clone MP16-2
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Enzyme
Type Primary
Immunogen A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa345-360) from rat ODC
Isotype IgG1

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Ornithine Decarboxylase (ODC) Ab-1, Mouse Monoclonal Antibody.

ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.

Host Species: Mouse

Clone: MP16-2

Isotype: IgG1

Species Reactivity: Human, Rat, and Mouse. Because epitope of MAb MP16-2 (-IWGPTC-) is present in all known eukaryotic ODC sequences, it should react with ODC from other eukaryotes also.

Epitope: aa 355-360 (-IWGPTC-). It is associated with active site of ODC or a locus in its direct vicinity.

Immunogen: A synthetic hexadecapeptide (KPDEKYYSSSIWGPTC) (aa 345-360) from rat ODC

Molecular Weight: 53kDa

Positive Control: Epithelial cells in normal placenta or prostate CA

Cellular Localization: Cytoplasmic Ab-1 recognizes the native ODC and not the inhibited (labeled with H3-DFMO) enzyme.

Recommended for:

  • Immunohistochemistry (Frozen only)
  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO