missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ormetoprim, TCI America™

Manufacturer:  TCI America O04241G

 View more versions of this product

Catalog No. O04241G



Specifications

Specifications

Ormetoprim
233°C
6981-18-6
MFCD00057747
KEEYRKYKLYARHO-UHFFFAOYSA-N
5-[(4,5-dimethoxy-2-methylphenyl)methyl]pyrimidine-2,4-diamine
23418
Crystal-Powder at 20°C
White-Yellow
1g
C14H18N4O2
2,4-Diamino-5-(4,5-dimethoxy-2-methylbenzyl)pyrimidine
CC1=CC(=C(C=C1CC2=CN=C(N=C2N)N)OC)OC
274.324
274.32
≥98.0% (HPLC,T)

Chemical Identifiers

6981-18-6
274.324
KEEYRKYKLYARHO-UHFFFAOYSA-N
23418
CC1=CC(=C(C=C1CC2=CN=C(N=C2N)N)OC)OC
C14H18N4O2
MFCD00057747
2,4-Diamino-5-(4,5-dimethoxy-2-methylbenzyl)pyrimidine
5-[(4,5-dimethoxy-2-methylphenyl)methyl]pyrimidine-2,4-diamine
Safety and Handling

Safety and Handling

TSCA : No

Documents