Please login to your online account to display your discounted pricing

Orlistat, Tocris™

Pancreatic, gastric and carboxylester lipase inhibitor; antiobesity and antihypercholesterolemic activity

$73.75 - $237.35

Chemical Identifiers

Chemical Name or Material Orlistat
Assay Percent Range >99%
Molecular Formula C29H53NO5
Formula Weight 495.73
Synonym N-Formyl-L-leucine (1S)-1-[[(2S, 3S)-3-hexyl-4-oxo-2-oxetanyl]methyl]dodecyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

354010

 
r & d systems
RDS354010
10mg Each for $73.75

354050

 
r & d systems
RDS354050
50mg Each for $237.35
Description & Specifications

Specifications

Formula Weight 495.73
Quantity 10mg
Synonym N-Formyl-L-leucine (1S)-1-[[(2S, 3S)-3-hexyl-4-oxo-2-oxetanyl]methyl]dodecyl ester
Molecular Formula C29H53NO5
Chemical Name or Material Orlistat
Assay Percent Range >99%
CAS 96829-58-2
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C