missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Orlistat, 98%

$99.47 - $442.59

Chemical Identifiers

CAS 96829-58-2
Molecular Formula C29H53NO5
Molecular Weight (g/mol) 495.745
MDL Number MFCD05662360
InChI Key AHLBNYSZXLDEJQ-FWEHEUNISA-N
Synonym orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
PubChem CID 3034010
IUPAC Name [(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl] (2S)-2-formamido-4-methylpentanoate
SMILES CCCCCCCCCCCC(CC1C(C(=O)O1)CCCCCC)OC(=O)C(CC(C)C)NC=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62999MF View Documents Alfa Aesar™
J62999MF
50mg Each for $99.47
 
AAJ62999MC View Documents Alfa Aesar™
J62999MC
100mg Each for $186.59
 
AAJ62999MD View Documents Alfa Aesar™
J62999MD
250mg Each for $442.59
 
Description

Description

Applications
Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase. More detailed experiments show that gradually THL affects the retinoblastoma protein pathway and leads to cell-cycle arrest at G1/S boundary. Lipase Inhibitor, THL is an inhibitor of pancreatic and gastric lipase, as well as carboxyl ester lipase (CEL) and diacylglycerol lipase-α (DAGLα). Lipase Inhibitor, THL also activates caspase-3.

Solubility
Soluble in DMSO (19 mg/mL), ethanol (20 mg/mL), and DMF (20 mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

96829-58-2
495.745
AHLBNYSZXLDEJQ-FWEHEUNISA-N
3034010
CCCCCCCCCCCC(CC1C(C(=O)O1)CCCCCC)OC(=O)C(CC(C)C)NC=O
C29H53NO5
MFCD05662360
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
[(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl] (2S)-2-formamido-4-methylpentanoate

Specifications

White
50mg
C29H53NO5
14,6869
AHLBNYSZXLDEJQ-FWEHEUNISA-N
[(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl] (2S)-2-formamido-4-methylpentanoate
3034010
Solid
Orlistat
<50°C
96829-58-2
MFCD05662360
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
CCCCCCCCCCCC(CC1C(C(=O)O1)CCCCCC)OC(=O)C(CC(C)C)NC=O
495.745
495.73
98%
Safety and Handling

Safety and Handling

RTECSNumber : OH3167600

Recommended Storage : −30° to −10°C