missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Orlistat, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals J62999MF

 View more versions of this product

Catalog No. AAJ62999MF


Description

Description

Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase. More detailed experiments show that gradually THL affects the retinoblastoma protein pathway and leads to cell-cycle arrest at G1/S boundary. Lipase Inhibitor, THL is an inhibitor of pancreatic and gastric lipase, as well as carboxyl ester lipase (CEL) and diacylglycerol lipase-α (DAGLα). Lipase Inhibitor, THL also activates caspase-3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Orlistat, used in obesity research, is a pancreatic lipase inhibitor that acts locally in the gastrointestinal tract to inhibit lipase. More detailed experiments show that gradually THL affects the retinoblastoma protein pathway and leads to cell-cycle arrest at G1/S boundary. Lipase Inhibitor, THL is an inhibitor of pancreatic and gastric lipase, as well as carboxyl ester lipase (CEL) and diacylglycerol lipase-α (DAGLα). Lipase Inhibitor, THL also activates caspase-3.

Solubility
Soluble in DMSO (19 mg/mL), ethanol (20 mg/mL), and DMF (20 mg/mL).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.
Specifications

Specifications

Orlistat
<50°C
C29H53NO5
50 mg
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
AHLBNYSZXLDEJQ-FWEHEUNISA-N
(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl (2S)-2-formamido-4-methylpentanoate
3034010
98%
96829-58-2
White
MFCD05662360
14,6869
Soluble in DMSO (19mg/mL),ethanol (20mg/mL),and DMF (20mg/mL).
CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O
495.75
495.73
Solid

Chemical Identifiers

96829-58-2
495.75
AHLBNYSZXLDEJQ-FWEHEUNISA-N
3034010
CCCCCCCCCCC[C@@H](C[C@@H]1OC(=O)[C@H]1CCCCCC)OC(=O)[C@H](CC(C)C)NC=O
C29H53NO5
MFCD05662360
orlistat, tetrahydrolipstatin, xenical, alli, orlipastat, --tetrahydrolipstatin, orlipastatum, orlipastatum inn-latin, thlp, tetrahydrolipastatin
(2S)-1-[(2S,3S)-3-hexyl-4-oxooxetan-2-yl]tridecan-2-yl (2S)-2-formamido-4-methylpentanoate
Safety and Handling

Safety and Handling

RTECSNumber : OH3167600

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only