Thermo Scientific™ Orion Star™ A112 Conductivity Benchtop Meter
Take simple conductivity, TDS and temperature measurements with the Thermo Scientific™ Orion™ Star A112 Conductivity Benchtop Meter with easy operation for basic lab testing
Manufacturer:  Thermo Scientific™ ORISTARA1120
 View more versions of this product
Catalog No. 13-645-507
Description
Description
Take simple, routine conductivity, total dissolved solids (TDS) and temperature measurements with the Thermo Scientific™ Orion™ Star A112 Conductivity Benchtop Meter, an easy-to-use and budget-friendly instrument for basic conductivity analysis in the lab. Obtain temperature compensated readings using linear option with 20 or 25 °C reference temperatures. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage.
Thermo Scientific Orion Star A112 Conductivity Benchtop Meters combine simplicity with accuracy. A large LCD displays conductivity or TDS readings along with temperature. Icons provide quick updates on battery life and completed calibration status. Simple button layout and onscreen messages help with calibration and setup menu choices.
Features and Benefits
  • Large, informative screen clearly shows the main information, including conductivity or TDS reading, temperature, meter mode and power source
  • Don't miss a reading—AUTOREAD™ locks in the stable reading on your screen and ready indicator alerts when readings are stable Selectable cell constant
  • Â Selectable reading reference temperatures of 20°C or 25°C for an accurate result
  • Included electrode arm and newly-designed holder make it easier to maintain and place probes into samples
  • Plug the meter into almost any power outlet with the included universal power adapter, or use four AA batteries (sold separately) to run the meter on DC power
  • Non-volatile memory holds up to 50 data points
  • The IP54-rated housing can handle splashes and can be wall-mounted if table space is a problem
  • 3 year meter replacement warranty
Specifications
Specifications
Orion Star A112 conductivity/TDS benchtop meter with stand
0.01 µS/cm to 200.0 mS/cm
0.5% of reading ±1 digit
1mg/L
Linear (0.02 to 9.99)
2-cell or 4-cell conductivity sensors with built-in temperature
20, 25°C
-5.0° to 105.0° C, 22.0° to 221.0° F
±0.1°C
1 point temperature offset calibration
LCD
Conductivity or TDS with temperature
Internal data point memory
Automatic data logging with Auto-Read measure mode; manual data logging with Continuous measure mode
English
None
Universal AC adapter (included) or 4 AA batteries (sold separately)
Four AA (sold separately)
IP-54
41° to 113°F (ambient)
5 to 85 %, non-condensing
2 lb.
1
Orion Star A112 Conductivity/TDS/Temperature Benchtop Meter
0.01 μS minimum, auto ranging up to 3 significant digits
1 to 19999mg/L
0.5% of reading ±1 digit
0.001 to 10cm-1
Linear
Conductivity and temperature
0.1°C, 0.1°F
Manual or automatic with ATC temperature probe
1
Simple with highlighted measure key, scroll and shortcut keys
Smart Stability and Smart Averaging
50
Non-volatile memory preserves log and settings
8 pin MiniDIN (conductivity sensor with built-in temperature)
Conductivity sensor (purchase separately)
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
2000 hours
CE, TUV 3-1, FCC Class A
5° to 45° C (ambient)
24cm x 18cm x 11cm (9.5 in. x 7.1 in. x 4.3 in.)
0.9kg
3 year meter warranty
Documents