missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ Standard All-in-One™ pH Buffer Kit, PROMO

Work with ease using this color-coded pH buffer kit that includes three buffers, storage solution and a storage bottle.

Manufacturer:  Thermo Scientific™ ORI910199PM

Catalog No. 13624500PM



Description

Description

  • pH buffers with temperature tables on the labels for accurate calibrations and NIST™ traceability
  • Manufactured under ISO 9000 quality standards
  • Color coded buffers: pH 4.01 buffer is pink; pH 7.00 buffer is yellow; and pH 10.01 buffer is blue
  • Includes: One 1 pint (475 mL) bottle each of pH 4.00 buffer (910104), pH 7.00 buffer (910107), pH 10.01 buffer (910110) and standard pH electrode storage solution (910001) and one pH electrode storage bottle.
Specifications

Specifications

for accurate calibrations and NISTΓäó traceability
Standard All-in-One pH Buffer Kit
See individual components
475mL
Storage Solution
Yes
pH 4.01, 7.00 and 10.01 buffers, storage solution
Documents
Promotions

Promotions

Limited warranty period: