Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes for Orion Series Meters
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes.
Manufacturer:  Thermo Scientific™ ORI8157BNURCA
 View more versions of this product
Catalog No. 13-642-676
Description
Description
Measure biological media, pharmaceuticals, foods and more with Thermo Scientific™ Orion™ ROSS Ultra™ Refillable pH/ATC Triode™ Combination Electrodes. They are ideal for any application where the use of metallic reference electrodes would contaminate the samples. ROSS Ultra pH/ATC electrodes offer more flexibility, accuracy and precision for your pH measurement needs. With built-in temperature sensors, these electrodes offer a convenient solution for your routine sample measurements. They have great stability and drift and come with a two-year replacement warranty.
Specifications
Specifications
Orion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and RCA Connector and 1 meter cable
2 years
0.01
0° to 100°C
Glass fiber
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
12mm
BNC Waterproof, RCA
ROSS
Mettler Toledo Five and Seven Series pH Meters
0 to 14
0.03pH for buffers using automatic temperature compensation
Rugged epoxy body
7
120mm
1m
Mettler Toledo Five and Seven series pH meters
pH/ATC Combination Electrode
Documents