Thermo Scientific™ Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes
No more waiting for pH readings to stabilize when measuring samples of varying temperatures.
Manufacturer:  Thermo Scientific™ ORI8302BNURCA
 View more versions of this product
Catalog No. 13-642-677
Description
Description
No more waiting for pH readings to stabilize when measuring samples of varying temperatures. Our Thermo Scientific™ Orion™ ROSS Ultra™ Triode™ pH electrodes combine the stability, precision and speed of response of the ROSS Ultra line with integrated temperature measurement. Orion ROSS electrodes contain no Silver or Silver Chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.
Specifications
Specifications
Orion ROSS Ultra Glass Triode pH/ATC Combination Electrodes with a BNC and RCA Connector and 1 meter cable
2 years
0.01
0° to 100°C
Ceramic
ROSS electrode 3 M KCl fill solution (Cat. No. 810007)
12mm
BNC Waterproof, RCA
ROSS
Mettler Toledo Five and Seven Series pH Meters
0 to 14
0.03pH for buffers using automatic temperature compensation
Chemical resistant glass body
7
120mm
1m
Mettler Toledo Five and Seven series pH meters
pH/ATC Combination Electrode
Documents
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.