Thermo Scientific™ Orion™ PRT300 Ink-Based Printer
Use the Thermo Scientific™ Orion™ PRT300 Ink Printer with legacy Thermo Scientific™ Orion™ Aplus and PerpHecT meters and create printouts to meet GLP requirements
$1,726.64 - $1,726.64
Specifications
For Use With A Series and PerpHecT meters
Recommended Accessories Replacement ink ribbon (Cat. No. PRT302)
Type Meter Accessories
View More Specs
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products ${productFamilyLength}
Catalog Number Mfr. No. Voltage Price Quantity  
Catalog Number Mfr. No. Voltage Price Quantity  
13-642-200 Thermo Scientific™
ORIPRT300
110VAC Each for $1,726.64
Specifications
Description
Use the Thermo Scientific™ Orion™ PRT300 Ink Printer with legacy Thermo Scientific™ Orion™ Aplus and PerpHecT meters including 115A+, 125A+, 150A+, 145A+, 162A, 250A+, 290A+, 410A+, 420A+, 520A+, 525A+, 710A+, 720A+, 805A+, 810A+, 850A+, 920A+, 330, 350, 370, 550A, 535A, 555A and 862A and create printouts to meet GLP requirements.
Includes:
  • Printer
  • Ribbon
  • Manual
  • Interface cable needed to connect the meter and printer
Recommended Accessories:
  • 1010009 Printer Paper
  • 1010005 Ink Cartridge
Ordering Information:
An optional computer interface cable 0ACBL0 is available for connecting meters to the serial port of an IBM or compatible PC. For other computers and communication devices, a suitable cable must be fabricated. The following technical information outlines the pin configurations, data transfer hardware and software settings for the 350 and 370 meters:
  • 1200 baud rate
  • Eight data bits
  • No parity
  • One start bit
  • One stop bit
Description & Specifications
Specifications
A Series and PerpHecT meters
Replacement ink ribbon (Cat. No. PRT302)
Meter Accessories