Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ pH Electrode Filling Solution


Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes.

Manufacturer: thermo scientific™  ORI610011

 View more versions of this product

 Disclaimers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 13-301-131

  • / Each
 

Description & Specifications

Specifications

Description Filling Solution for pHuture pH Electrodes
For Use With pH electrodes and reference electrodes, Only use ROSS filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
Packaging Type Bottle
Quantity 60mL
Solution Type Filling solution

Use Thermo Scientific™ Orion™ pH Electrode Filling Solution with pH electrodes and reference electrodes. Packaged as five 2 oz. (5 x 60mL) bottles.

  • Only use ROSS filling solution (810007) with ROSS™ and ROSS Ultra™ pH electrodes to prevent damaging the ROSS electrodes
  • When the single junction reference electrode (900100) or double junction reference electrode (900200) are used with Orion ion selective electrodes (ISE), refer to the recommended ISE filling solution listed in the ISE user guide

Note:

The following part number has been changed due to a recent system upgrade.

Old Part Number:910008

New Part Number: 910008-WA

Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Data Safety Sheet (MSDS) files.