Thermo Scientific™ Orion™ Nitrate Sure-Flow Combination Electrode PROMO
Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.
Manufacturer:  Thermo Scientific™ 9707BNWPPM
Catalog No. 13642270PM
Description
Description
  • Calibration standards catalog number: 920706 - 0.1m NaNO3; 920707 - 1000 ppm N; 930707 - 100 ppm N
  • Required ISA catalog number: 930711 or 930710 / nitrate ISS
Specifications
Specifications
ISEmeters
Combination Nitrate Electrode with Waterproof BNC Connector
Free-flowing Sure-FlowTM junction assures stable
Combination Ion Selective Electrode
dri ft.-free potentials