Thermo Scientific™ Orion™ Lead Electrodes
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes.
Manufacturer:  Thermo Scientific™ ORI9682BNWP
 View more versions of this product
Catalog No. 13-645-161
Description
Description
Measure lead ions in aqueous solutions quickly, simply, accurately and economically with Thermo Scientific™ Orion™ Lead Electrodes. No separate reference electrode is needed with this electrode, making it convenient to use with small sample volumes.
  • Free-flowing Themo Scientific™ Sure-Flow™ junction assures stable, drift-free potentials
  • When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap
  • Lead Half-Cell Electrode used in conjunction with the Orion 900200 Double Junction Reference electrode offers the benefits of the Sure-Flow™ junction design
  • Calibration standards catalog number: 948206 - 0.1 M Pb(CIO4)2; 948207 - 0.1 M Na2SO4
  • Required ISA Catalog numbers : Consult instruction manual
Specifications
Specifications
Lead Ion Selective Electrode, BNC waterproof connector
With waterproof BNC connector
Free-flowing Sure-Flow™ junction assures stable
Waterproof BNC
drift-free potentials
ISE Meters
Documents
Accessories
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.