Thermo Scientific™ Orion™ Dual Star™ pH, ISE, mV, ORP and Temperature Dual Channel Benchtop Meter PROMO
Accurately monitor pH, ion concentration, mV, ORP and temperature on two channels with the Thermo Scientific™ Orion™ Dual Star pH / ISE Benchtop Meter for advanced lab analysis.
Manufacturer:  Thermo Scientific™ ORI2115001SD
 View more versions of this product
Catalog No. 13642782SD
Description
Description
The Orion Dual Star pH/ISE dual channel benchtop meter accelerates sample measurement throughput by showing the results of two channels simultaneously on its large, backlit display. Measure pH, ion concentration (ISE), mV, relative mV or ORP with temperature on two channels using the meter’s two BNC, reference pin-tip and ATC temperature inputs. Collect critical measurements using an assortment of read modes and onscreen ready indicator that alerts when readings are stable. The meter’s Smart Stability and Smart Averaging functions automatically compensate for measurement conditions, so the meter displays more accurate results faster.
  • View large individual measurements on a full screen display or quickly toggle to a split screen display to view two measurements at the same time
  • Easy-to-follow onscreen text prompts for calibration and setup menus available in a multi-language user interface with English, Spanish, French, German or Italian options
  • EZ Startup menu facilitates quick and easy first time meter setup
  • Numeric keypad and menu-specific function keys allow for quick and efficient meter setup and data entry
  • Selectable read modes include Auto-Read, On-Ready, At Timed Intervals, Continuous, Value Change in Measurement and Timed for a variety of measurement collection methods for different analytical methods
  • Perform up to a 6 point pH calibration with automatic recognition for USA/NIST and EURO/DIN buffers with option to manually input custom buffer values
  • Perform up to a 6 point ion concentration (ISE) calibration with non-linear auto-blank and low-concentration stability options or use incremental methods for single/double known addition and single/double known subtraction
  • Non-volatile meter memory holds up to 1000 data points with time and date stamp
  • Easily transfer data via USB and RS232 to a printer or computer with free Orion Dual Star Com communication software
  • Stir samples with ease using the meter-controlled Orion Star stirrer probe
  • Meter-attachable electrode stand assists with moving and maintaining electrodes
  • IP54-rated housing handles splashes and is wall-mountable to save space
Warranty: Three-year meter replacement warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Orion Dual Star pH/ISE dual channel benchtop meter kit with ROSS Ultra glass-body pH electrode, ATC probe, solutions and stand
Dual Star Meter with Electrode Stand; Universal Power Adapter; 8102BNUWP ROSS Ultra Glass Refillable pH Electrode; 927007MD Stainless Steel ATC Probe; 810001 ROSS Storage Solution; 910104, 910107, 910110 pH 4.01, 7.00, 10.01 pH Buffers
±0.002pH
±1999.9mV
±1999.9mV
±0.2 mV or ±0.05% of reading whichever is greater
0.0001 minimum, 1 to 3 significant digits (user selectable)
Segmented (point-to-point) slope, non-linear selectable auto-blank, low concentration range stability, incremental techniques including single known addition, single known subtraction, double known addition and double known subtraction
1 to 6
1 point relative mV (RmV) mode, 1 point ORP mode
-5.0° to 105.0°C, 22.0° to 221.0°F
±0.1°C
1 point temperature offset calibration
Yes, adjustable
Numeric with menu-specific function keys and dual purpose scroll/shortcut keys
Smart Stability and Smart Averaging
Automatic data logging with Auto-Read, On Ready, At Timed Intervals, Value Change In Measurement and Timed Reading measure modes; manual data logging with Continuous measure mode
High/low limit alarm
10 per channel with password protection
USB, RS232
pH electrode, ORP electrode, ion selective electrode (ISE), reference half-cell electrode, ATC temperature probe, stirrer probe (includes 8102BNUWP ROSS Ultra pH electrode, 927007MD Orion ATC probe)
Universal AC adapter (included)
90 to 260VAC 50/60Hz
5° to 45°C (ambient)
24 x 17 x 9cm (9.5 x 6.7 x 3.5 in.)
0.9kg
3 year meter warranty
Orion Dual Star pH/ISE/mV/RmV/ORP/Temperature Dual Channel Benchtop Meter
0.1, 0.01, 0.001pH
0.1, 0.01, 0.001pH
±1999.9mV
0.1mV
0.0001 to 19900 with selectable units of ppm, Molar, mg/L, % or unit-less
±0.2 mV or ±0.05% of reading whichever is greater
pH with calibration editing option, ion concentration (ISE) with calibration editing option, relative mV (RmV), ORP and temperature
2 to 6
7.000pH (adjustable), 1.000 ISE (adjustable)
0.1°C, 0.1°F
Manual or automatic with ATC temperature probe
2
Graphic LCD with backlight
pH, mV, relative mV (RmV), ORP or ion concentration (ISE) with temperature
Transfer single, range or all data points to printer or computer
1000 with date and time stamp
Non-volatile memory preserves data log, calibration log and meter settings
Two BNC (pH, ORP or ion selective electrode), two pin-tip (reference electrode), two 8 pin MiniDIN (ATC temperature probe), stirrer input (stirrer probe)
English, Spanish, French, Italian and German
Universal 50-60 Hz, 100-240 VAC power adapter (Cat. No. 1010003)
90 to 260VAC
41° to 113°F (ambient)
5 to 85 %, non-condensing
2 lb.
CE, TUV 3-1, FCC Class A
Documents
Promotions
Promotions
  • Thermo Scientific
    Get 30% off select Thermo Scientific™ lab equipment, including incubators, ovens, mixers, meters, and more.
    09/30/18