Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ Cadmium Electrodes
Measure cadmium ions in aqueous solutions quickly, simply, accurately and economically.
$1,822.61 - $1,822.61
Specifications
Junction Free-flowing Sure-Flow junction assures stable, drift-free potentials
View More Specs
 Disclaimers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Connector Types Price Quantity    
13-645-159
thermo scientific™
ORI9648BNWP
Waterproof BNC Each for $1,822.61
Description & Specifications
Specifications
Junction Free-flowing Sure-Flow junction assures stable, drift-free potentials
Measure Cadmium ions in aqueous solutions quickly, simply and economically with the Thermo Scientific™ Orion™ Cadmium Electrodes.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small sample volumes.
Related Products
  • Calibration standards: Consult instruction manual
  • Required ISA: 940011
Warranty: 12 months from date of purchase by Buyer or 18 months from date of shipment from Seller, whichever is earlier.