missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes

Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions by using these pH combination electrodes.

Manufacturer:  Thermo Scientific™ ORI9135APWP

 View more versions of this product

Catalog No. 13-641-216



Description

Description

Read viscous liquid samples quickly and accurately without the sample coming into contact with silver ions with Thermo Scientific™ Orion™ AquaPro™ pH Combination Electrodes, which have a clog-resistant and open double junction reference system. These electrodes are ideal for measurements in samples containing proteins or TRIS buffers and that do not contain mercury. The electrodes require minimal maintenance because there is no fill solution; instead they have a polymer gel reference.

Feature improved electrode life, low maintenance, fast response time and clean junctions. Patented reference system and double-junction design prevent silver ions from contacting sample.
  • Measure 0 to 14pH
  • Temperature range: 0° to 60°C (0° to 140°F)
  • Choice of connectors with 1m (3ft.) cable
Specifications

Specifications

Epoxy Body
0 to 14
0° to 60°C
120mm
Waterproof BNC
12mm
4.72 in.
32° to 140°F
Combination Ag/AgCl double juction pH Electrode, BNC waterproof connoector
Orion pH Meter
±0.02pH
Open
1m
0.47 in.
Ag/AgCl Double Junction
39 in.
Moist surfaces, solids, semisolids, small sample sizes
Always store in pH storage solution, Orion 910001. Either store in a beaker of storage solution or reassemble in the storage bottle the electrode was provided with.
Documents

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.