Thermo Scientific™ Orion™ AQUAfast Colorimetric Auto-Test Ampoule Reagents, Ammonia - HR
Minimize the risks of inaccurate measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials.
Manufacturer:  Thermo Scientific™ AC4011
Catalog No. 36-099-3818
Description
Description
Minimize the risks of improper measurement and contamination with Thermo Scientific™ Orion™ AQUAfast™ IV Chemistry Reagents, which are contained in self-filling vials.
  • No. of test: 30
  • Consistency and reproducibility improved with AQUAfast ampoules and an AQ4000 meter
  • Thermo Scientific™ Auto-ID™ eliminates measurement errors