Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ 9120APWP KNIpHE™ pH Electrode

Perform accurate determination of pH in meat matrices with the Thermo Scientific™ Orion™ 9120APWP KNIpHE™ pH Electrode.

$453.37 - $453.37

Specifications

Connector Types BNC
Junction Ceramic
Filling Solution Type 3M KCl
Internal Reference Ag/AgCl Double Junction
pH Range 0 to 14
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Description Price Quantity    

13-303-159

 
thermo scientific™
ORI9121APWP
Orion 9121APWP Double Junction Replacement KNIpHE Combination pH electrode Each for $453.37
Description & Specifications

Specifications

Connector Types BNC
Junction Ceramic
Filling Solution Type 3M KCl
Internal Reference Ag/AgCl Double Junction
pH Range 0 to 14
Storage Requirements Store electrode in Orion pH electrode storage solution 910001
Temperature (English) Operating 32° to 140°F
Temperature (Metric) Operating 0° to 60°C

Perform accurate determination of pH in meat matrices with the Thermo Scientific™ Orion™ 9120APWP KNIpHE™ pH Electrode. It eliminates breakage with a stainless steel cutting blade surrounding a special combination pH electrode.

Ordering Information:

  • 9120APWP

Includes:

  • Stainless steel cutting blade
  • pH electrode
  • Electrode housing
  • 60 ml bottle of electrode fill solution