Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode

Measure general pH samples as well as viscous or colloidal samples with the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.

$977.44 - $977.44

Specifications

Applications Biological/Pharmaceutical –TRIS buffer, proteins, enzymes; Emulsions – Foods, cosmetics, oils; Emulsions – Petroleum products, paint; General Purpose – Most sample types; High Ionic Strength – Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength –
Description Orion ROSS Sure-Flow pH electrode
Diameter (Metric) Cap 16mm
Connector Types Waterproof BNC
Material Glass body
View More Specs

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Products
Catalog Number Mfr. No. Connector Types Price Quantity    

13-642-243

  Accessories
thermo scientific™
ORI8172BNWP
Waterproof BNC Each for $977.44
Description & Specifications

Specifications

Applications Biological/Pharmaceutical –TRIS buffer, proteins, enzymes; Emulsions – Foods, cosmetics, oils; Emulsions – Petroleum products, paint; General Purpose – Most sample types; High Ionic Strength – Acids, bases, brines, pH > 12 or pH < 2; Low Ionic Strength –
Description Orion ROSS Sure-Flow pH electrode
Diameter (Metric) Cap 16mm
Connector Types Waterproof BNC
Material Glass body
Filling Solution Type 3M KCl (810007)
Includes Detachable bulb guard, 2 x 60mL bottles of ORion ROSS electrode fill solution
Internal Reference ROSS
pH Range 0 to 14
Precision 0.01
Temperature (Metric) Operating 0° to 100°C
Type Glass body

Measure general pH samples as well as viscous or colloidal samples with the free-flowing sleeve junction design of the Thermo Scientific™ Orion™ 8172BNWP ROSS™ Sure-Flow™ pH Electrode.

This general purpose pH electrode offers superior performance for dirty, difficult samples such as soils, sludges, colloids, viscous materials and organics. Junction cleans easily between measurements; pressing down on the cap allows filling solution to flow through the junction carries away any contamination that may interfere with measurement.

  • Material: Glass body
  • pH range: 0 to 14
  • Temperature range: 0° to 100°C
  • Internal reference: ROSS
  • Junction: Sure-Flow
  • Dimensions: 120 × 12mm
  • pH precision: 0.01
  • Fill solution: 3M KCl, Orion Cat. No 810007
  • Cap diameter: 16mm

Includes:

  • Detachable bulb guard, two bottles of Orion ROSS electrode fill solution