Please login to your online account to display your discounted pricing
Thermo Scientific™ Orion™ 815600 ROSS™ Combination pH Electrode
Calculate results three to five times more precise than those obtained with conventional electrodes with the Thermo Scientific™ Orion™ 815600 ROSS™ Combination pH Electrode.
$771.18 - $771.18
Specifications
Drift <0.002pH/day
Junction Glass Fiber
Material Epoxy body
Filling Solution Type 3M KCl
Internal Reference ROSS
View More Specs
Products
Catalog Number Mfr. No. Connector Types Price Quantity    
13-641-7
thermo scientific™
ORI8155SC
S7 screw cap Each for $771.18
Description & Specifications
Specifications
Drift <0.002pH/day
Junction Glass Fiber
Material Epoxy body
Filling Solution Type 3M KCl
Internal Reference ROSS
pH Range 0 to 14
Precision 0.01
Temperature (English) Operating 32° to 212°F
Temperature (Metric) Operating 0° to 100°C
Calculate results three to five times more precise than those obtained with conventional electrodes with the Thermo Scientific™ Orion™ 815600 ROSS™ Combination pH Electrode. This electrode provides stable readings to 0.01 pH in less than 30 seconds.
Features
  • For general purpose when precise pH determination is required
  • For work in extreme case of samples varying from one another by 50°C or more
  • Epoxy body for ruggedness and durability
  • Drift is less than 0.002 pH per day; restandardization is minimized