Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ 266S Advanced Waterproof Portable pH/mV Meter

Measure and log pH, mV and temperature using the Thermo Scientific™ Orion™ 266S Intrinsically Safe (US) Portable pH Meter

$4,186.05 - $4,186.05

Specifications

Accuracy (mV) 0.1% of reading ±0.3 mV
Accuracy (pH) ±0.01pH
Accuracy (Temperature) ±0.3°C
Backlight Option None
Battery Life 1000 hours
View More Specs
Products
Catalog Number Mfr. No. Description Includes Price Quantity    

13-302-111

 
thermo scientific™
ORI0266S3
Orion Intrinsically Safe 266S pH portable meter kit with sensor, field case, beakers and solutions Orion 266S pH meter, 9109WP Orion Triode gel-filled epoxy-body pH/ATC electrode with 1.5 meter cable, field case, beakers, pH buffers, 3 AA batteries Each for $4,186.05
Description & Specifications

Specifications

Accuracy (mV) 0.1% of reading ±0.3 mV
Accuracy (pH) ±0.01pH
Accuracy (Temperature) ±0.3°C
Backlight Option None
Battery Life 1000 hours
Battery Type Three AA (included)
Calibration pH
Calibration Points (pH) 1 to 3
Certifications/Compliance CE, FM (IS, Class I Div 1 Group A, B, C, D, T4), Ex (Intrinsically Safe)
Channels 1
Data Management Details Transfer single or all data points to printer or computer
Data Points 100 with time and date stamp
Display Type LCD
Dimensions (L x W x H) 16cm x 13.3cm x 3cm (6.30 in. x 5.24 in. x 1.18 in.)
IP Rating IP-66
Languages English
Log Function Type Automatic by interval or event or manual data logging
Outputs RS232
Range (mV) ±1300.0mV
Range (pH) -2.00 to 16.00pH
Resolution (pH) 0.1, 0.01pH
Inputs E-DIN Waterproof (pH electrode), banana plug (ATC temperature probe)
Keypad Measure, cal, print, scroll up, scroll down, STO, RCL, clock, power keys
Measurement Modes pH with mV and temperature
Model Orion Intrinsically Safe 266S pH/mV/Temperature Portable Meter
Power Supply 3 AA batteries (included)
Range (Temperature) -20° to 120 °C, -4° to 248° F
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Temperature (English) Operating 14° to 131°F (ambient)
Temperature (Metric) Operating -10° to 55°C (ambient)
Temperature Selection Automatic with ATC temperature probe
Warranty 3 year meter warranty
Weight (English) 1.4 lb.
Weight (Metric) 0.6kg

Measure pH, mV and temperature in environments where non-sparking equipment is required using the Thermo Scientific™ Orion™ 266S Intrinsically Safe (US) Portable pH Meter. Log up to 100 data points and use the RS232 port to transfer data to an external device. Intrinsically safe approval rating by Factory Mutual (FM) for US only.

Features:

  • Meter can be controlled by a remote PC via RS-232-C output and software for transferring data.
  • Compact, portable waterproof meter simultaneously shows pH and temperature on watertight LCD.
  • Meter features mV mode, GLPcheck™ electrode verification and three-point autocalibration.
  • Three AA alkaline batteries supply power for up to 2000 hours of continuous use.
  • Meter can be controlled by a remote PC via RS-232-C output and software for transferring data to the computer.

Includes:

  • Meter Only package includes meter, built-in quiver, neckstrap and instruction manual, RS232 cable and software
  • Meter/Electrode package includes meter, Triode™ 3-in-1 Combination pH Electrode, built-in quiver, neckstrap and instruction manual
  • Meter Field Kit package includes meter; built-in quiver; neckstrap and instruction manual; case; stand; electrode holder; beakers; 4, 7, 10 buffers; RS232 cable and software

Compatible with:

0900A0 Orion Model 900A Printer