Please login to your online account to display your discounted pricing

Thermo Scientific™ Orion™ 261S Basic Waterproof Portable pH Meter

Measure pH, mV and temperature using the Thermo Scientific™ Orion™ 261S Intrinsically Safe (US) Portable pH Meter

$2,909.69 - $2,909.69

Specifications

Accuracy (mV) 0.1% of reading ±0.3 mV
Accuracy (pH) ±0.01pH
Accuracy (Temperature) ±0.3°C
Backlight Option None
Battery Life 1000 hours
View More Specs
Products
Catalog Number Mfr. No. Description Price Quantity    

13-302-114

 
thermo scientific™
ORI0261S2
Orion Intrinsically Safe 261S pH portable meter Each for $2,909.69
Description & Specifications

Specifications

Accuracy (mV) 0.1% of reading ±0.3 mV
Accuracy (pH) ±0.01pH
Accuracy (Temperature) ±0.3°C
Backlight Option None
Battery Life 1000 hours
Battery Type Three AA (included)
Calibration pH
Calibration Points (pH) 1 to 3
Certifications/Compliance CE, FM (IS, Class I Div 1 Group A, B, C, D, T4), Ex (Intrinsically Safe)
Channels 1
Display Type LCD
Dimensions (L x W x H) 16cm x 13.3cm x 3cm (6.30 in. x 5.24 in. x 1.18 in.)
IP Rating IP-66
Languages English
Outputs None
Range (mV) ±1300.0mV
Range (pH) -2.00 to 16.00pH
Resolution (pH) 0.1, 0.01pH
Inputs E-DIN Waterproof (pH electrode), banana plug (ATC temperature probe)
Keypad Measure, cal, scroll up, scroll down, power keys
Measurement Modes pH with mV and temperature
Model Orion Intrinsically Safe 261S pH/mV/Temperature Portable Meter
Power Supply 3 AA batteries (included)
Range (Temperature) -20° to 120 °C, -4° to 248° F
Resolution (mV) 0.1mV
Resolution (Temperature) 0.1°C
Temperature (English) Operating 14° to 131°F (ambient)
Temperature (Metric) Operating -10° to 55°C (ambient)
Temperature Selection Automatic with ATC temperature probe
Warranty 3 year meter warranty
Weight (English) 1.4 lb.
Weight (Metric) 0.6kg

Measure pH, mV and temperature in environments where non-sparking equipment is required using the Thermo Scientific™ Orion™ 261S Intrinsically Safe (US) Portable pH Meter. Intrinsically safe approval rating by Factory Mutual (FM) for US only.

Features:

  • 2.00 to 16.00pH range
  • -20° to +120° temperature range
  • ±1300mV range
  • Safely measure in hazardous and confined space environments and anywhere nonsparking equipment is desired
  • Battery-operated
  • Large, easy-to-read display to monitor electrode condition, calibration status and pH, mV and temperature readings
  • Automatic or manual calibration with up to three calibration points

Includes:

  • Meter Only package includes meter, built-in quiver, neckstrap and instruction manual
  • Meter/Electrode package includes meter, Triode™ 3-in-1 Combination pH Electrode, built-in quiver, neckstrapand instruction manual
  • Meter Field Kit package includes meter, built-in quiver, neckstrapand instruction manual, case, stand, electrode holder, beakers, 4, 7, 10 buffers, RS232 cable and software