Please login to your online account to display your discounted pricing

Organometallic Oil Standard, 21-Element, 100μg/g, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 100g
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  S21100Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. S21-100Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
Chemical Name or Material 21 Element Oil Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Nonhazardous
Solvent or Matrix Base Oil 75
Purity Grade Oil
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition (by Analyte or Components) Each component at 100μg/g (S21-100), 300μg/g (S21-300), 500μg/g (S21-500), or 900μg/g (S21-900) in Base Oil 75:
Aluminum
Barium
Boron
Cadmium
Calcium
Chromium
Copper
Iron
Lead
Magnesium
Manganese
Molybdenum
Nickel
Phosphorous
Silicon
Silver
Sodium
Tin
Titanium
Vanadium
Zinc

Specifications

Chemical Name or Material 21 Element Oil Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/g
Al100μg/g
B100μg/g
Ba100μg/g
Ca100μg/g
Cd100μg/g
Cr100μg/g
Cu100μg/g
Fe100μg/g
Mg100μg/g
Mn100μg/g
Mo100μg/g
Na100μg/g
Ni100μg/g
P100μg/g
Pb100μg/g
Si100μg/g
Sn100μg/g
Ti100μg/g
V100μg/g
Zn100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Ambient