missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Organometallic Oil Standard, 21-Element, 300μg/g, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$480.14 - $749.37

Specifications

Chemical Name or Material 21 Element Oil Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S21-300Y
View Documents
Spex Certiprep Llc
S21300Y
100g Triple-leached LDPE bottle
Each for $749.37
 
S21-300Z
View Documents
Spex Certiprep Llc
S21300Z
50g Triple-leached LDPE bottle
Each for $480.14
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag300μg/g
Al300μg/g
B300μg/g
Ba300μg/g
Ca300μg/g
Cd300μg/g
Cr300μg/g
Cu300μg/g
Fe300μg/g
Mg300μg/g
Mn300μg/g
Mo300μg/g
Na300μg/g
Ni300μg/g
P300μg/g
Pb300μg/g
Si300μg/g
Sn300μg/g
Ti300μg/g
V300μg/g
Zn300μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

21 Element Oil Standard
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
SDS
Product Certifications

Product intended for laboratory use only