missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Organometallic Oil Standard, 12-Element, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Manufacturer:  Spex Certiprep Llc S12100Y

 View more versions of this product

Catalog No. S12-100Y


 

Disclaimers:

Product intended for laboratory use only


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Specifications

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag100μg/g
Al100μg/g
Cr100μg/g
Cu100μg/g
Fe100μg/g
Mg100μg/g
Na100μg/g
Ni100μg/g
Pb100μg/g
Si100μg/g
Sn100μg/g
Ti100μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Description & Specifications

Specifications

12 Element Oil Standard
Each component at 100μg/g (S12-100), 300μg/g (S12-300), 500μg/g (S12-500), or 900μg/g (S12-900) in Base Oil 75:
Aluminum
Chromium
Copper
Iron
Lead
Magnesium
Nickel
Silicon
Silver
Sodium
Tin
Titanium
With Certificate of Analysis.
Base Oil 75
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Oil
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
100g

Chemical Identifiers

AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
12 Element Oil Standard

Safety and Handling

DOTInformation : Nonhazardous

Recommended Storage : Ambient