Please login to your online account to display your discounted pricing

Organochloride Pesticide Mixture, 2000μg/mL in Hexane/Toluene (1:1), Ultra Scientific


Quantity 1 x 1mL

Manufacturer: ultra scientific  US127

 View more versions of this product

Catalog No. US-US-127

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1 x 1mL
Physical Form Liquid
Name Note 2000μg/mL
Chemical Name or Material Organochloride Pesticide Mixture
CAS 000319-84-6
CAS 000319-86-8
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 000108-88-3
CAS 000319-85-7
CAS 000110-54-3
CAS 000309-00-2

Specifications

Chemical Name or Material Organochloride Pesticide Mixture
Name Note 2000μg/mL
CAS 000110-54-3
CAS 000108-88-3
CAS 000309-00-2
CAS 000319-84-6
CAS 000319-85-7
CAS 000319-86-8
Physical Form Liquid