Please login to your online account to display your discounted pricing

Organochloride Pesticide Mixture, 2000μg/mL in Hexane/Toluene (1:1), Ultra Scientific

$550.54 - $550.54

Chemical Identifiers

Chemical Name or Material Organochloride Pesticide Mixture
Name Note 2000μg/mL
CAS 000110-54-3
CAS 000108-88-3
CAS 000309-00-2
CAS 000319-84-6
CAS 000319-85-7
CAS 000319-86-8
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-US-127

 
ultra scientific
US127
1 x 1mL Each for $550.54
Description & Specifications

Specifications

Quantity 1 x 1mL
Physical Form Liquid
Name Note 2000μg/mL
Chemical Name or Material Organochloride Pesticide Mixture
CAS 000319-84-6
CAS 000319-86-8
Concentration or Composition (by Analyte or Components) 2000μg/mL
CAS 000108-88-3
CAS 000319-85-7
CAS 000110-54-3
CAS 000309-00-2