Please login to your online account to display your discounted pricing

Org 37684, Tocris™

5-HT2 agonist

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material Org 37684
Assay Percent Range >98%
Molecular Formula C14H19NO2·HCl
Formula Weight 269.77
Synonym (3S)-3-[(2, 3--Dihydro-5-methoxy-1H-inden-4-yl)oxy]pyrrolidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

479710

 
r & d systems
479710
10mg Each for $218.74

479750

 
r & d systems
479750
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 269.77
Quantity 10mg
Assay Percent Range >98%
CAS 213007-95-5
Synonym (3S)-3-[(2, 3--Dihydro-5-methoxy-1H-inden-4-yl)oxy]pyrrolidine hydrochloride
Chemical Name or Material Org 37684
Molecular Formula C14H19NO2·HCl
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature