missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Org 27569, Tocris

Potent allosteric CB1 modulator

$212.57 - $889.32

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C24H28ClN3O
Formula Weight 409.95
Synonym 5-Chloro-3-ethyl-N-[2-[4-(1-piperidinyl)phenyl]ethyl-1H-indole-2-carboxamide
Chemical Name or Material Org 27569
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
295710 R & D Systems
RDS295710
10mg Each for $212.57
295750 R & D Systems
RDS295750
50mg Each for $889.32
Specifications Description & Specifications

Chemical Identifiers

>99%
409.95
Org 27569
C24H28ClN3O
5-Chloro-3-ethyl-N-[2-[4-(1-piperidinyl)phenyl]ethyl-1H-indole-2-carboxamide

Specifications

10mg
>99%
409.95
Org 27569
868273-06-7
C24H28ClN3O
5-Chloro-3-ethyl-N-[2-[4-(1-piperidinyl)phenyl]ethyl-1H-indole-2-carboxamide

Safety and Handling

Recommended Storage : Store at Room Temperature