missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Org 25543 hydrochloride, Tocris™

Potent and selective GlyT2 inhibitor

$257.85 - $1,073.10

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C24H32lN2O4·HCl
Formula Weight 448.98
Synonym N-[[1-(Dimethylamino)cyclopentyl]methyl]-3, 5-dimethoxy-4-(phenylmethoxy)benzamide hydrochloride
Chemical Name or Material Org 25543 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
478210 R & D Systems
RDS478210
10mg Each for $257.85
478250 R & D Systems
RDS478250
50mg Each for $1,073.10
Specifications Description & Specifications

Chemical Identifiers

>99%
448.98
Org 25543 hydrochloride
C24H32lN2O4·HCl
N-[[1-(Dimethylamino)cyclopentyl]methyl]-3, 5-dimethoxy-4-(phenylmethoxy)benzamide hydrochloride

Specifications

10mg
>99%
448.98
Org 25543 hydrochloride
495076-64-7
C24H32lN2O4·HCl
N-[[1-(Dimethylamino)cyclopentyl]methyl]-3, 5-dimethoxy-4-(phenylmethoxy)benzamide hydrochloride

Safety and Handling

Recommended Storage : Desiccate at Room Temperature