Please login to your online account to display your discounted pricing

Org 24598 lithium salt, Tocris™

Selective GlyT1 inhibitor

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material Org 24598 lithium salt
Assay Percent Range >98%
Molecular Formula C19H19F3LiNO3
Formula Weight 373.3
Synonym N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine lithium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

403510

 
r & d systems
RDS403510
10mg Each for $244.81

403550

 
r & d systems
RDS403550
50mg Each for $997.23
Description & Specifications

Specifications

Formula Weight 373.3
Quantity 10mg
Chemical Name or Material Org 24598 lithium salt
Assay Percent Range >98%
Molecular Formula C19H19F3LiNO3
CAS 722456-08-8
Synonym N-Methyl-N-[(3R)-3-phenyl-3-[4-(trifluoromethyl)phenoxy]propyl]-glycine lithium salt
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature