missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Org 12962 hydrochloride

Selective 5-HT2C agonist

$199.70 - $834.54

Specifications

Product Type Org 12962 hydrochloride
Inhibitors Org 12962 hydrochloride
Molecular Weight 302.12
Purity >99
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
358510R Tocris™
358510
10mg Each for $199.70
358550R Tocris™
358550
50mg Each for $834.54
Specifications

Description

Selective 5-HT2C receptor agonist (pEC50 values are 7.01, 6.38 and 6.28 for 5-HT2C, 5-HT2A and 5-HT2B respectively). Displays antiaversive effects in a rat model of panic-like anxiety.
  • Chemical Name: 1-[6-Chloro-5-trifluoromethyl)-2-pyridinyl]-piperazine hydrochloride
  • Formula: C10H11ClF3N3.HCl
  • CAS Number: 210821-63-9
  • Solubility: Soluble to 50mM in water and to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

Org 12962 hydrochloride
Org 12962 hydrochloride
302.12
>99