Please login to your online account to display your discounted pricing

Alfa Aesar™ Orexin B, Mouse, Rat

For cellular and molecular biology applications

Specifications

Product Type Orexin B, mouse, rat
Cross Reactivity Mouse, Rat
Molecular Weight 2935.38
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Asn-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65275EXD

 
alfa aesar™
J65275EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ65275MCR

 
alfa aesar™
J65275MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Orexin B, mouse, rat
Cross Reactivity Mouse, Rat
Molecular Weight 2935.38
Storage Requirements -30° to -10°C
Sequence Arg-Pro-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Asn-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2
Format Solid
Regulatory Status RUO
Purification Method HPLC