Please login to your online account to display your discounted pricing

Alfa Aesar™ Orexin B, Human

For cellular and molecular biology applications

$238.59 - $238.59

Specifications

Product Type Orexin B, human
Cross Reactivity Human
Molecular Weight 2899.33
Storage Requirements -30° to -10°C
Sequence Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65110EXD

 
alfa aesar™
J65110EXD
0.5mg Each for $238.59

AAJ65110MCR

 
alfa aesar™
J65110MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Orexin B, human
Cross Reactivity Human
Molecular Weight 2899.33
Storage Requirements -30° to -10°C
Sequence Arg-Ser-Gly-Pro-Pro-Gly-Leu-Gln-Gly-Arg-Leu-Gln-Arg-Leu-Leu-Gln-Ala-Ser-Gly-Asn-His-Ala-Ala-Gly-Ile-Leu-Thr-Met-NH2
Format Lyophilized powder
Regulatory Status RUO
Purification Method HPLC