Please login to your online account to display your discounted pricing

Orcinol Monohydrate, 99%, ACROS Organics™

$392.35 - $392.35

Chemical Identifiers

Chemical Name or Material Orcinol monohydrate
CAS 6153-39-5
Assay ≥98.5% (HPLC)
Assay Percent Range 99%
Molecular Formula C7H8O2·H2O
Formula Weight 142.15
MDL Number MFCD00149092
Synonym 3, 5-Dihydroxytoluene monohydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC129551000

  SDS
fisher scientific acros
129551000
Glass bottle 100g Each for $392.35

AC129555000

  SDS
thermo fisher scientific
AC129555000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (HPLC)
Physical Form Powder, Crystals or Flakes
Formula Weight 142.15
Melting Point 56° to 61°C
Quantity 100g
Color White to off-white
Beilstein 06, 882
Merck Index 14, 6864
Solubility Solubility in water: soluble in water. Other solubilities: soluble in alcohol and ether, freely soluble in benzene, slightly soluble in chloroform and carbon, disulfide
Water 10 to 14% (K.F.)
Infrared Spectrum Authentic
Molecular Formula C7H8O2·H2O
MDL Number MFCD00149092
CAS 6153-39-5
Packaging Glass bottle
Synonym 3, 5-Dihydroxytoluene monohydrate
Chemical Name or Material Orcinol monohydrate
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.