missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Orange G, Electrophoresis Grade

Collagen stain for connective tissue

$17.42 - $198.93

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H12N2Na2O7S2
Molecular Weight (g/mol) 454.379
MDL Number MFCD00012457
InChI Key UFUQLGWUULNYBM-DDVLFWKVSA-N
Synonym C.I. 16230; Acid Orange 10
PubChem CID 131850556
IUPAC Name (8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6274314 View Documents Alfa Aesar™
J6274314
25g Each for $17.42
 
AAJ6274322 View Documents Alfa Aesar™
J6274322
100g Each for $46.12
 
AAJ62743A1 View Documents Alfa Aesar™
J62743A1
1kg Each for $198.93
 
Specifications

Chemical Identifiers

1936-15-8
454.379
UFUQLGWUULNYBM-DDVLFWKVSA-N
131850556
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
C16H12N2Na2O7S2
MFCD00012457
C.I. 16230; Acid Orange 10
(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium

Specifications

1936-15-8
MFCD00012457
C.I. 16230; Acid Orange 10
UFUQLGWUULNYBM-DDVLFWKVSA-N
(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
131850556
Powder
Red to Orange
Orange G
C16H12N2Na2O7S2
4120705
Soluble in water or methyl cellosolve; Slightly soluble in ethanol
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
454.379
452.38
Electrophoresis
25g
Safety and Handling

Safety and Handling

RTECSNumber : QJ6500000

TSCA : TSCA