missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Orange G, 85+%, pure, high purity biological stain, ACROS Organics™

$66.87 - $194.06

Chemical Identifiers

CAS 1936-15-8
Molecular Formula C16H12N2Na2O7S2
Molecular Weight (g/mol) 454.379
MDL Number MFCD00012457
InChI Key UFUQLGWUULNYBM-DDVLFWKVSA-N
Synonym 7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic acid, disodium salt, Acid Orange 10, C.I. 16230
PubChem CID 131850556
IUPAC Name (8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229820250
SDS View Documents
Fisher Scientific Acros
229820250
25g Glass bottle
Each for $66.87
 
AC229821000
SDS View Documents
Thermo Fisher Scientific
229821000
100g Glass bottle
Each for $194.06
 
Description

Description

Specifications

Chemical Identifiers

1936-15-8
454.379
UFUQLGWUULNYBM-DDVLFWKVSA-N
131850556
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
C16H12N2Na2O7S2
MFCD00012457
7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic acid, disodium salt, Acid Orange 10, C.I. 16230
(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium

Specifications

1936-15-8
85+%
MFCD00012457
7-Hydroxy-8-(phenylazo)-1, 3-naphthalenedisulfonic acid, disodium salt, Acid Orange 10, C.I. 16230
Authentic
(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonic acid;sodium
131850556
Powder
Glass bottle
Orange to Orange-Red
Orange G
85.0% min.
C16H12N2Na2O7S2
16, 300
UFUQLGWUULNYBM-DDVLFWKVSA-N
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
454.379
452.35
Pure
Complies
25g
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS