Please login to your online account to display your discounted pricing

Optovin, Tocris™

Reversible photoactive TRPA1 activator

$212.74 - $886.68

Chemical Identifiers

Chemical Name or Material Optovin
Assay Percent Range >99%
Molecular Formula C15H13N3OS2
Formula Weight 315.41
Synonym 5-[[2, 5-Dimethyl-1-(3-pyridinyl)-1H-pyrrol-3-yl]methylene]-2-thioxo-4-thiazolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

490150

 
r & d systems
RDS490150
50mg Each for $886.68

490110

 
r & d systems
RDS490110
10mg Each for $212.74
Description & Specifications

Specifications

Formula Weight 315.41
Quantity 50mg
Chemical Name or Material Optovin
Molecular Formula C15H13N3OS2
Synonym 5-[[2, 5-Dimethyl-1-(3-pyridinyl)-1H-pyrrol-3-yl]methylene]-2-thioxo-4-thiazolidinone
Recommended Storage Store at +4°C
Assay Percent Range >99%
CAS 348575-88-2
Safety and Handling

Store at +4°C