Please login to your online account to display your discounted pricing

OPTIZYME™ XbaI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80085

 View more versions of this product

Catalog No. BP8008-5

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
For Use With (Application) Indicates ligation efficiency of DNA fragments generated by digestion with the Restriction Enzyme
Quantity 10000U
Includes 10X OPTIZYME™ Buffer 4
Format Liquid
Product Name XbaI
Storage Buffer 10mM Tris HCl (pH 7.4), 100mM KCl, 7mM 2-mercaptoethano, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Concentration 50U/μL
Units 10,000U
pH 7.4
Cut Site T.CTAGA