Please login to your online account to display your discounted pricing

OPTIZYME™ T4 Polynucleotide Kinase, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80981

 View more versions of this product

Catalog No. BP80981

  • / Each
 

Description & Specifications

Specifications

Product Name T4 Polynucleotide Kinase
Concentration 10U/μL
Source E. coli cells with a cloned pse T gene of bacteriophage T4
pH 7.5
Quantity 500U
For Use With (Application) Probes for hybridization or transcript mapping, primers for PCR or DNA sequencing and markers for Gel electrophoresis, removal of 3'-phosphate groups
Storage Buffer 10X OPTIZYME™ T4 PNK Buffer 1 (for forward reaction): 500mM Tris HCl (7.6 pH at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine, 10X OPTIZYME™ T4 PNK Buffer 2 (for exchange reaction): 500mM imidazole-HCl
Storage Requirements -20°C
Format Liquid
Concentration 10U/μL
Quantity 500U
Color Colorless
Format Liquid