Please login to your online account to display your discounted pricing

OPTIZYME™ T4 DNA Polymerase, Fisher BioReagents™


Manufacturer: fisher scientific company  BP81055

 View more versions of this product

Catalog No. BP81055

  • / Each
 

Description & Specifications

Specifications

pH 7.5
Quantity 500U
Source E. coli cells with a cloned gene 43 of bacteriophage T4
Storage Buffer OPTIZYME™ T4 DNA Pol Buffer 5X: Tris HCl 335mM (8.8 pH at 25°C), MgCl2 33mM, DTT 5mM, (NH4)2SO4 84mM
Format Liquid
Concentration 5U/μL
Color Colorless
For Use With (Application) Synthesis of labeled DNA probes, creation of blunt PCR products with 3'-dA overhangs, ligation-independent cloning of PCR products, Restriction Enzymes, reverse transcriptases and T4 DNA ligase
Product Name T4 DNA Polymerase
Storage Requirements -20°C