Please login to your online account to display your discounted pricing

OPTIZYME™ T4 DNA Ligase, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80991

 View more versions of this product

Catalog No. BP80991

  • / Each
 

Description & Specifications

Specifications

Source E. coli cells with a cloned gene 30 of bacteriophage T4
Storage Requirements -20°C
Color Colorless
Quantity 100U
Storage Buffer OPTIZYME™ T4 DNA Ligase Buffer 10X: Tris HCl 400mM, MgCl2 100mM, DTT 100mM, ATP 5mM (7.8 pH at 25°C), OPTIZYME™ PEG 4000 Solution 50%: Polyethylene Glycol 50% (w/v)
Product Name T4 DNA Ligase
For Use With (Application) Used to clone restriction Enzyme generated DNA fragments, cloning PCR products, ligation of double-stranded Oligonucleotide linkers or adaptors to DNA, site-directed mutagenesis, amplified fragment length polymorphism, ligase-mediated RNA detection, nick
pH 7.5
Concentration 5U/μL
Format Liquid