Please login to your online account to display your discounted pricing

OPTIZYME™ SmaI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80115

 View more versions of this product

Catalog No. BP8011-5

  • / Each
 

Description & Specifications

Specifications

Units 5,000U
pH 7.4
Cut Site CCC.GGG
Includes 10X OPTIZYME™ Buffer 4
Incubator Temperature 30°C
Concentration 10U/μL
Format Liquid
For Use With (Application) >90% of pUC19 DNA fragments can be ligated and re-cut after 50-fold over-digestion with SmaI
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
Product Name SmaI
Quantity 5000U
Format Liquid
Applications >90% of pUC19 DNA fragments can be ligated and re-cut after 50-fold over-digestion with SmaI
Product Name SmaI
Quantity 5000U