Please login to your online account to display your discounted pricing

OPTIZYME™ PvuII, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80225

 View more versions of this product

Catalog No. BP8022-5

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Storage Requirements Keep container tightly closed
Includes >50% Glycerin, 25 to 50% Water
Incubator Temperature 37°C
Units 5,000U
pH 7.4
Cut Site CAG.CTG
Concentration 10U/μL
For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with Pvu II
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Quantity 5000U
Product Name PvuII
Quantity 5000U
Product Name PvuII