Please login to your online account to display your discounted pricing

OPTIZYME™ NotI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80041

 View more versions of this product

Catalog No. BP8004-1

  • / Each
 

Description & Specifications

Specifications

Storage Buffer 20mM Tris HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
Storage Requirements Keep container tightly closed
Format Liquid
Product Name NotI
Concentration 10U/μL
pH 7.4
Quantity 300U
Units 300U
Includes 25 to 50% Water, >50% Glycerin
Cut Site GC.GGCCGC
For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Not I