Please login to your online account to display your discounted pricing

OPTIZYME™ NcoI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80175

 View more versions of this product

Catalog No. BP8017-5

  • / Each
 

Description & Specifications

Specifications

Product Name NcoI
pH 7.4
Quantity 2500U
Units 2,500U
For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nco I
Storage Requirements Keep container tightly closed
Format Liquid
Concentration 10U/μL
Includes 25 to 50% Water, >50% Glycerin
Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Cut Site C.CATGG
Applications >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Nco I
Format Liquid
Product Name NcoI
Quantity 2500U